Learn More
3-(2,3-Dichlorobenzoyl)-propionic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(2,3-Dichlorobenzoyl)-propionic Acid |
| CAS | 32003-41-1 |
| Molecular Formula | C10H8Cl2O3 |
| Quantity | 100 mg |
| Synonym | 2,3-Dichloro-gamma-oxo-benzenebutanoic Acid |
| InChI Formula | InChI=1S/C10H8Cl2O3/c11-7-3-1-2-6(10(7)12)8(13)4-5-9(14)15/h1-3H,4-5H2,(H,14,15) |
| SMILES | OC(=O)CCC(=O)c1cccc(Cl)c1Cl |
| IUPAC Name | 4-(2,3-dichlorophenyl)-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 247.07 |
| Formula Weight | 245.985 |