Learn More
3-(1-Cyanoethyl)benzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-[(1RS)-1-Cyanoethyl]benzoic Acid |
| CAS | 5537-71-3 |
| Molecular Formula | C10 H9 N O2 |
| Quantity | 10 g |
| Synonym | 3-[(1RS)-1-Cyanoethyl]benzoic acid, Ketoprofen Imp. G (EP), Benzoic acid, 3-(1-cyanoethyl)-, Benzoic acid, m-(1-cyanoethyl)- (7CI,8CI), 3-(1-Cyanoethyl)benzoic acid, 2-(3-Carboxyphenyl)propionitrile, DF 2107Y, NSC 113992, m-(1-Cyanoethyl)benzoic acid |
| InChI Formula | InChI=1S/C10H9NO2/c1-7(6-11)8-3-2-4-9(5-8)10(12)13/h2-5,7H,1H3,(H,12,13) |
| SMILES | CC(C#N)c1cccc(c1)C(=O)O |
| IUPAC Name | 3-(1-cyanoethyl)benzoic acid |
| Molecular Weight (g/mol) | 175.18 |
| Formula Weight | 175.0633 |