missing translation for 'onlineSavingsMsg'
Learn More

2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents

Product Code. 11386445
Click to view available options
Quantity:
10,000 g
100 g
2000 g
500 g
Packaging Type:
Poly Bottle
Poly Pail
Unit Size:
100g
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11386445

Brand: Fisher BioReagents BP246610

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Additive 2xyt Agar
Form Powder
For Use With (Application) Molecular Genetics
Packaging Type Poly Pail
Quantity 10,000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.