Learn More
(2S,3S,5S)-2-Amino-3-hydroxy-5-(tert-butyloxycarbonylamino)-1,6-diphenylhexane, TRC
Specifications
Specifications
| Chemical Name or Material | (2S,3S,5S)-2-Amino-3-hydroxy-5-(tert-butyloxycarbonylamino)- |
| CAS | 144163-85-9 |
| Molecular Formula | C23 H32 N2 O3 |
| Quantity | 25 g |
| Synonym | Ritonavir Impurity 24 |
| InChI Formula | InChI=1S/C23H32N2O3/c1-23(2,3)28-22(27)25-19(14-17-10-6-4-7-11-17)16-21(26)20(24)15-18-12-8-5-9-13-18/h4-13,19-21,26H,14-16,24H2,1-3H3,(H,25,27)/t19-,20-,21-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@H](C[C@H](O)[C@@H](N)Cc1ccccc1)Cc2ccccc2 |
| IUPAC Name | tert-butyl N-[(2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl]carbamate |
| Molecular Weight (g/mol) | 384.51 |
| Formula Weight | 384.2413 |