Learn More
(2S,3R)-3-Hydroxynorleucine, TRC
Specifications
Specifications
| Chemical Name or Material | (2S,3R)-3-Hydroxynorleucine |
| CAS | 10148-69-3 |
| Molecular Formula | C6H13NO3 |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C6H13NO3/c1-2-3-4(8)5(7)6(9)10/h4-5,8H,2-3,7H2,1H3,(H,9,10)/t4-,5+/m1/s1 |
| SMILES | N[C@H](C(O)=O)[C@H](O)CCC |
| IUPAC Name | (2S,3R)-2-amino-3-hydroxyhexanoic acid |
| Molecular Weight (g/mol) | 147.17 |
| Formula Weight | 147.0895 |