missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™

Product Code. p-3770950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10284784 1 g Glass bottle 1g
10244834 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10284784 Supplier Thermo Scientific Acros Supplier No. 395010010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 98760-08-8
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.34
MDL Number MFCD00671705,MFCD02258997
InChI Key NVPOUMXZERMIJK-UHFFFAOYNA-N
Synonym 2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
PubChem CID 9813904
IUPAC Name tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: <0.1%
Formula Weight 263.34
Percent Purity 98%
Chemical Name or Material (2R, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.