Learn More
(2R,3R,5S)-3,5-Dimethyl-1-benzyloxy-2-hydroxy-8-(4-methoxyphenoxy)-4-octanone, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,3R,5S)-3,5-Dimethyl-1-benzyloxy-2-hydroxy-8-(4-methoxyphenoxy)-4-octanone |
| CAS | 276690-15-4 |
| Molecular Formula | C24H32O5 |
| Quantity | 1 mg |
| Synonym | (2R,3R,5S)-2-Hydroxy-8-(4-methoxyphenoxy)-3,5-dimethyl-1-(phenylmethoxy)-4-octanone |
| InChI Formula | InChI=1S/C24H32O5/c1-18(8-7-15-29-22-13-11-21(27-3)12-14-22)24(26)19(2)23(25)17-28-16-20-9-5-4-6-10-20/h4-6,9-14,18-19,23,25H,7-8,15-17H2,1-3H3/t18?,19?,23-/m0/s1 |
| SMILES | O=C([C@H](C)[C@@H](O)COCC1=CC=CC=C1)[C@@H](C)CCCOC2=CC=C(OC)C=C2 |
| IUPAC Name | (2R,3R,5S)-1-(benzyloxy)-2-hydroxy-8-(4-methoxyphenoxy)-3,5-dimethyloctan-4-one |
| Molecular Weight (g/mol) | 400.51 |
| Formula Weight | 400.22 |