Learn More
(2R)-3-[4-(carbamoylamino)phenyl]-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2R)-3-[4-(carbamoylamino)phenyl]-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic Acid |
| CAS | 324017-22-3 |
| Molecular Formula | C25H23N3O5 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C25H23N3O5/c26-24(31)27-16-11-9-15(10-12-16)13-22(23(29)30)28-25(32)33-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,28,32)(H,29,30)(H3,26,27,31)/t22-/m1/s1 |
| SMILES | NC(=O)Nc1ccc(C[C@@H](NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O)cc1 |
| IUPAC Name | (2R)-3-[4-(carbamoylamino)phenyl]-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
| Molecular Weight (g/mol) | 445.47 |
| Formula Weight | 445.1638 |