Learn More
KGW Isotherm™ 2700mL Dish-Shaped Dewar Flasks
Click to view available options
Capacity (Metric):
2700 mL
Unit Size:
Each
Spezifikation
Spezifikation
| Diameter (Metric) Outer | 230mm |
| For Use With (Application) | Cooling of small samples |
| Height (Metric) Inner | 125mm |
| Height (Metric) | 160mm |
| Capacity (Metric) | 2700 mL |
| Diameter (Metric) Inner | 200 mm |
| Material | Borosilicate Glass |