missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ASP/C3a desArg Protein

Product Code. 18781809
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18781809

Brand: R&D Systems™ 9620C3025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce IL-6 secretion by LPS-stimulated human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 0.4-2.4 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01024
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 718
Molecular Weight (g/mol) M.W. (Observed): 9 kDa, reducing conditions; M.W. (theoretical): 9 kDa
Name SKINT2
Quantity 25 μg
Source E. coli-derived human ASP/C3a desArg protein Ser672-Ala747
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Common Name SKINT2
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ASP/C3a desArg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.