Learn More
MedChemExpress Cyclo(his-pro)
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| CAS | 53109-32-3 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 234.25 g/mol |
| Product Type | Natural Products |
| Quantity | 25 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : ≥ 30 mg/mL (128.07 mM) |
| Synonym | Cyclo(histidyl-proline)∣ Histidylproline diketopiperazine |