missing translation for 'onlineSavingsMsg'
Learn More

Acepromazine Maleate, TRC

Product Code. 30133666
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133666 25 mg 25mg
30133726 50 mg 50mg
17913495 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133666 Supplier Toronto Research Chemicals Supplier No. TRCA13090025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3598-37-6
Molecular Formula C19 H22 N2 O S . C4 H4 O4
Molecular Weight (g/mol) 442.53
Synonym Acepromazine Maleate, Ethanone, 1-[10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:1), Ethanone, 1-[10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, (Z)-2-butenedioate (1:1), Ketone, 10-[3-(dimethylamino)propyl]phenothiazin-2-yl methyl, maleate (1:1) (8CI), Ketone, 10-[3-(dimethylamino)propyl]phenothiazin-2-yl methyl, maleate (6CI,7CI), Maleic acid, compd. with 10-(3-dimethylaminopropyl)phenothiazin-2-yl methyl ketone (6CI), Acepran, Acepromazine monomaleate, Acetopromazine maleate salt, Acetylpromazine maleate, Anatran, Atravet, Calmivet, Calmo Neosan, NSC 264522, Notensil, Notenzil, Plegicil, Sedalin, Soprontin
IUPAC Name 1-[10-[3-(dimethylamino)propyl]phenothiazin-2-yl]ethanone
SMILES CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(=O)C.OC(=O)\C=C/C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H22N2OS/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)23-19/h4-5,7-10,13H,6,11-12H2,1-3H3
Formula Weight 442.1562
Percent Purity >95%
Chemical Name or Material Acepromazine Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.