Learn More
Fisher Chemical 25KG CALCIUM CHLORIDE FUSED25kg
Brand:  Fisher Chemical C/1320/70
25KG CALCIUM CHLORIDE FUSED25kg
Product Code. 10654242
  • ONLINE EXCLUSIVE PRICE
    422.00 €
    413.00 € / 25kg
    Save 9.00 €
    2% Off
Please to purchase this item. Need a web account? Register with us today!
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Calcium chloride fused
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Calcium chloride