Learn More
Ibidi 25 micro-Inserts 4 Well FulTrac for Self-insertion
Description
- 25 x 4 well silicone insert used for tracing individual cells (adherent and suspension)
- in a 10 cm transport dish
- sterilized
Specifications
Specifications
| Type | Culture Insert |
| Quantity | 25 |
| Sterility | Sterile |
| No. of Wells | 4 |