missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibidi 25 Culture-Inserts 3 Well for Self-insertion
Click to view available options
No. of Wells:
3
Unit Size:
Pack of 25
Description
- 25 x 3 well silicone insert with 2 defined cell-free gaps of 500 μm
- suitable for wound healing
- migration assays
- 2D invasion assays
- and co-cultivation of cells
- in a 10 cm transport dish
- sterilized
Specifications
Specifications
| Type | Culture Insert |
| Quantity | 25 |
| Sterility | Sterile |
| No. of Wells | 3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction