Learn More
24-Epicerebrosterol, TRC
Specifications
Specifications
| Chemical Name or Material | 24-Epicerebrosterol |
| CAS | 27460-26-0 |
| Molecular Formula | C27H46O2 |
| Quantity | 10 mg |
| Synonym | (24R)-Cholest-5-ene-3beta,24-diol, (24R)-Hydroxycholesterol |
| InChI Formula | InChI=1S/C27H46O2/c1-17(2)25(29)11-6-18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20+,21+,22-,23+,24+,25-,26+,27-/m1/s1 |
| SMILES | CC(C)[C@H](O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-hydroxy-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 402.65 |
| Formula Weight | 402.3498 |