Learn More
22-Keto-21-thia-3,6,9-trioxatricosan-1-ol, TRC
Product Code. 30406221
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406221 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30406221 Supplier Toronto Research Chemicals Supplier No. TRCK31500010MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 22-Keto-21-thia-3,6,9-trioxatricosan-1-ol
CAS 130727-50-3
Molecular Formula C26 H52 O5 S
Quantity 10 mg
InChI Formula InChI=1S/C26H52O5S/c1-2-3-4-5-10-13-16-23-32-24-17-14-11-8-6-7-9-12-15-18-29-19-20-30-21-22-31-26(28)25-27/h27H,2-25H2,1H3
SMILES CCCCCCCCCSCCCCCCCCCCCOCCOCCOC(=O)CO
IUPAC Name 2-[2-(11-nonylsulfanylundecoxy)ethoxy]ethyl 2-hydroxyacetate
Molecular Weight (g/mol) 476.75
Formula Weight 476.3535