Learn More
21-Deacetoxy 11-Oxodeflazacort, TRC
Specifications
Specifications
| Chemical Name or Material | 21-Deacetoxy 11-Oxodeflazacort |
| CAS | 13649-83-7 |
| Molecular Formula | C23H27NO4 |
| Quantity | 50 mg |
| Synonym | 2'-Methyl-5'betaH-pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione |
| InChI Formula | InChI=1S/C23H27NO4/c1-12(25)23-19(28-13(2)24-23)10-17-16-6-5-14-9-15(26)7-8-21(14,3)20(16)18(27)11-22(17,23)4/h7-9,16-17,19-20H,5-6,10-11H2,1-4H3/t16-,17-,19+,20+,21-,22-,23+/m0/s1 |
| SMILES | CC(=O)[C@@]12N=C(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4C(=O)C[C@]23C |
| IUPAC Name | (1S,2S,4R,8S,9S,12S,13R)-8-acetyl-6,9,13-trimethyl-5-oxa-7-azapentacyclo[10.8.0.02,9.04,8.013,18]icosa-6,14,17-triene-11,16-dione |
| Molecular Weight (g/mol) | 381.46 |
| Formula Weight | 381.194 |