Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ R-A6x250/8MI Fixed Rotors
Product Code. p-7158842 Shop All Ohaus Products
Click to view available options
Capacity:
1 x 50 mL (29 mm dia.)
1 x 50 mL (30 mm dia)
2 x 30 mL
4 x 15 mL
5 x 10 mL
8 x 1.5 mL
Unit Size:
Pack of 2
Product Code. 15886642
Brand: Ohaus™ 30304369
Please to purchase this item. Need a web account? Register with us today!
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-A6x250/8MI Fixed Rotors
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-A6x250/8MI Fixed Rotors
TRUSTED_SUSTAINABILITY
Specifications
Capacity 8 x 1.5 mL
Max. Tube Diameter 11 mm
For Use With (Equipment) R-A6x250/8MI Fixed Rotors
Description Adapter, 8x1.5ml D11mm, 2/pk
No. of Places 8
No. per Pack 2