missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis

Product Code. 11309156
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309156 1 g 1g
11329156 250 mg 250mg
11319156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309156 Supplier Thermo Scientific Alfa Aesar Supplier No. L20280.03
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 745783-91-9 | C9H5F3O2 | 202.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745783-91-9
Molecular Formula C9H5F3O2
Molecular Weight (g/mol) 202.13
MDL Number MFCD08533288
InChI Key HBOSPPHIPGYHDI-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylglyoxal hydrate, 2-oxo-2-2-trifluoromethyl phenyl acetaldehyde hydrate, 2-oxo-2-2-trifluoromethyl phenyl ethanal, oxamethane, benzeneacetaldehyde, a-oxo-2-trifluoromethyl-,hydrate 1:1
PubChem CID 2783281
IUPAC Name 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde;hydrate
SMILES FC(F)(F)C1=CC=CC=C1C(=O)C=O

Specifications

Melting Point 36°C to 43°C
Quantity 1 g
Formula Weight 202.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 2-(Trifluoromethyl)phenylglyoxal hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.