Learn More
2-(Trifluoromethyl)benzamide, 98+%, Thermo Scientific™
Product Code. 11428056
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11428056
Brand: Thermo Scientific Alfa Aesar A14103.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 360-64-5
Molecular Formula C8H6F3NO
Molecular Weight (g/mol) 189.14
MDL Number MFCD00014798
InChI Key QBAYIBZITZBSFO-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzamide, o-trifluoromethylbenzamide, o-trifluoromethyl benzamide, unii-5u8y7e6aio, 2-trifluoromehtyl benzamide, benzamide, 2-trifluoromethyl, o-trifluoromethyl-benzamide, 5u8y7e6aio, synquest 4848-3-12
PubChem CID 67759
IUPAC Name 2-(trifluoromethyl)benzamide
SMILES NC(=O)C1=CC=CC=C1C(F)(F)F
Specifications
Melting Point 161°C to 164°C
Quantity 5 g
Beilstein 2616148
Formula Weight 189.14
Percent Purity ≥98%
Chemical Name or Material 2-(Trifluoromethyl)benzamide