Learn More
2-Thiouridine 2’,3’,5’-Triacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Thiouridine 2',3',5'-Triacetate |
| CAS | 28542-31-6 |
| Molecular Formula | C15 H18 N2 O8 S |
| Quantity | 250 mg |
| Synonym | 2,3-Dihydro-2-thioxo-1-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)-4(1H)-pyrimidinone |
| InChI Formula | InChI=1S/C15H18N2O8S/c1-7(18)22-6-10-12(23-8(2)19)13(24-9(3)20)14(25-10)17-5-4-11(21)16-15(17)26/h4-5,10,12-14H,6H2,1-3H3,(H,16,21,26)/t10-,12-,13-,14-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=CC(=O)NC2=S |
| IUPAC Name | [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(4-oxo-2-sulfanylidenepyrimidin-1-yl)oxolan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 386.377 |
| Formula Weight | 386.078 |