Learn More
2-(tert-Butylamino)-4,6-dichloro-1,3,5-triazine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(tert-Butylamino)-4,6-dichloro-1,3,5-triazine |
| CAS | 27282-85-5 |
| Molecular Formula | C7 H10 Cl2 N4 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C7H10Cl2N4/c1-7(2,3)13-6-11-4(8)10-5(9)12-6/h1-3H3,(H,10,11,12,13) |
| SMILES | CC(C)(C)Nc1nc(Cl)nc(Cl)n1 |
| IUPAC Name | N-tert-butyl-4,6-dichloro-1,3,5-triazin-2-amine |
| Molecular Weight (g/mol) | 221.09 |
| Formula Weight | 220.0283 |