Learn More
2-[tert-Butyl(nitroso)amino]-1-(3-chlorophenyl)propan-1-one, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[tert-Butyl(nitroso)amino]-1-(3-chlorophenyl)propan-1-one |
| CAS | 2763780-10-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13H17ClN2O2 |
| Quantity | 250 mg |
| Synonym | 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)nitrosoamino]- (ACI), 1-(3-Chlorophenyl)-2-[(1,1-dimethylethyl)nitrosoamino]-1-propanone (ACI), N-Nitroso Bupropion |
| InChI Formula | InChI=1S/C13H17ClN2O2/c1-9(16(15-18)13(2,3)4)12(17)10-6-5-7-11(14)8-10/h5-9H,1-4H3 |
| SMILES | O=NN(C(C(=O)C=1C=CC=C(Cl)C1)C)C(C)(C)C |
| IUPAC Name | N-tert-butyl-N-[1-(3-chlorophenyl)-1-oxopropan-2-yl]nitrous amide |
| Molecular Weight (g/mol) | 268.74 |
| Show More |