Learn More
2-Quinoxalinecarboxylic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Quinoxaline-2-carboxylate |
| CAS | 1865-11-8 |
| Molecular Formula | C10 H8 N2 O2 |
| Quantity | 250 mg |
| Synonym | 2-Quinoxalinecarboxylic Acid Methyl Ester, Methyl Quinoxaline-2-carboxylate |
| InChI Formula | InChI=1S/C10H8N2O2/c1-14-10(13)9-6-11-7-4-2-3-5-8(7)12-9/h2-6H,1H3 |
| SMILES | COC(=O)c1cnc2ccccc2n1 |
| IUPAC Name | methyl quinoxaline-2-carboxylate |
| Molecular Weight (g/mol) | 188.18 |
| Formula Weight | 188.0586 |