Learn More
2-Pyridineacetonitrile, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Pyridineacetonitrile |
| CAS | 2739-97-1 |
| Molecular Formula | C7H6N2 |
| Quantity | 10 g |
| Synonym | (2-Pyridinyl)acetonitrile, (Pyridyl-2-yl)acetonitrile, 2-(2-Pyridyl)acetonitrile |
| InChI Formula | InChI=1S/C7H6N2/c8-5-4-7-3-1-2-6-9-7/h1-3,6H,4H2 |
| SMILES | N#CCC1=CC=CC=N1 |
| IUPAC Name | 2-(pyridin-2-yl)acetonitrile |
| Molecular Weight (g/mol) | 118.14 |
| Formula Weight | 118.05 |