missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridineacetamide, 98%, Thermo Scientific™

Product Code. 11451457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451457 1 g 1g
11461457 5 g 5g
11471457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451457 Supplier Thermo Scientific Alfa Aesar Supplier No. B24939.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Pyridineacetamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5451-39-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00087532
InChI Key UXVCEKRAZBZVSL-UHFFFAOYSA-N
Synonym pyridine-2-acetamide, 2-pyridineacetamide, 2-pyridin-2-yl acetamide, 2-2-pyridyl acetamide, 2-pyridyl acetamide, acmc-20ajt2, 2-2-pyridinyl acetamide, 2-pyridine acetamide, 2-pyridine-2-yl-acetamide
PubChem CID 227129
IUPAC Name 2-pyridin-2-ylacetamide
SMILES NC(=O)CC1=CC=CC=N1

Specifications

Melting Point 118°C to 120°C
Quantity 1 g
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 2-Pyridineacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.