missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridin-3-ylaniline, 95%, Thermo Scientific™

Product Code. 10422124
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10422124

Brand: Thermo Scientific Maybridge CC41814DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 177202-83-4
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.22
MDL Number MFCD08060516
InChI Key MTYHJHATMPCNLW-UHFFFAOYSA-N
Synonym 2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine
PubChem CID 459519
SMILES NC1=CC=CC=C1C1=CN=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name 2-(pyridin-3-yl)aniline
Formula Weight 170.21
Percent Purity 95%
Chemical Name or Material 2-Pyridin-3-ylaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.