Learn More
2-Propylhexanoic Acid, TRC
Product Code. 30419248
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419248 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30419248 Supplier Toronto Research Chemicals Supplier No. TRCP83710025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Propylhexanoic Acid
CAS 3274-28-0
Molecular Formula C9 H18 O2
Quantity 25 mg
Synonym 2-Propylhexanoic Acid, 4-Octanecarboxylic acid
InChI Formula InChI=1S/C9H18O2/c1-3-5-7-8(6-4-2)9(10)11/h8H,3-7H2,1-2H3,(H,10,11)
SMILES CCCCC(CCC)C(=O)O
IUPAC Name 2-propylhexanoic acid
Molecular Weight (g/mol) 158.24
Formula Weight 158.1307