Learn More
2-Propylhexanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Propylhexanoic Acid |
| CAS | 3274-28-0 |
| Molecular Formula | C9 H18 O2 |
| Quantity | 1 g |
| Synonym | 2-Propylhexanoic Acid, 4-Octanecarboxylic acid |
| InChI Formula | InChI=1S/C9H18O2/c1-3-5-7-8(6-4-2)9(10)11/h8H,3-7H2,1-2H3,(H,10,11) |
| SMILES | CCCCC(CCC)C(=O)O |
| IUPAC Name | 2-propylhexanoic acid |
| Molecular Weight (g/mol) | 158.24 |
| Formula Weight | 158.1307 |