missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol-d{8}, 99%(Isotopic)

Product Code. 11354368
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354368 1 g 1g
11374368 5 g 5g
11364368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11354368 Supplier Thermo Scientific Alfa Aesar Supplier No. 042314.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 22739-76-0 | C3H8O | 68.15 g/mol

2-Propanol-d{8} is used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22739-76-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 68.15
MDL Number MFCD00044341
InChI Key KFZMGEQAYNKOFK-PIODKIDGSA-N
Synonym 2-propanol-d8, isopropanol-d8, 2 h? propan-2-2 h ol, octadeuteroisopropanol, 2h8-2-propanol, 2-propan-1,1,1,2,3,3,3-d7-ol-d, 2-propanol-d isotopic 1g, 2-propanol-d8, 99.5 atom % d
PubChem CID 2723972
SMILES [2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Density 0.90g/mL
Flash Point −11.7°C (53°F)
Odor Alcohol-like
Quantity 1 g
Melting Point -89.5°C
Boiling Point 82.4°C
UN Number UN1219
IUPAC Name 1,1,1,2,3,3,3-heptadeuterio-2-deuteriooxypropane
Formula Weight 68.15
Percent Purity 99%
Chemical Name or Material 2-Propanol-d8, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.