Learn More
2-(prop-2-yn-1-yl)-1,2,3,4-tetrahydroisoquinoline-1,3-dione, TRC
Product Code. 30336778
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336778 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336778 Supplier Toronto Research Chemicals Supplier No. TRCB52643525MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(prop-2-yn-1-yl)-1,2,3,4-tetrahydroisoquinoline-1,3-dione
CAS 410538-30-6
Molecular Formula C12H9NO2
Quantity 25 mg
InChI Formula InChI=1S/C12H9NO2/c1-2-7-13-11(14)8-9-5-3-4-6-10(9)12(13)15/h1,3-6H,7-8H2
SMILES O=C1Cc2ccccc2C(=O)N1CC#C
IUPAC Name 2-prop-2-ynyl-4H-isoquinoline-1,3-dione
Molecular Weight (g/mol) 199.21
Formula Weight 199.0633