Learn More
2-Picolinic-d4 Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Picolinic-d4 Acid |
| CAS | 284487-61-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 D4 H N O2 |
| Quantity | 5 mg |
| Synonym | 2-Pyridine-3,4,5,6-d4-carboxylic acid (9CI), 2-Picolinic-d4 Acid, 2-Picolinic Acid-D4 |
| InChI Formula | InChI=1S/C6H5NO2/c8-6(9)5-3-1-2-4-7-5/h1-4H,(H,8,9)/i1D,2D,3D,4D |
| SMILES | [2H]c1nc(C(=O)O)c([2H])c([2H])c1[2H] |
| IUPAC Name | 3,4,5,6-tetradeuteriopyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 127.134 |
| Show More |