Learn More
2-Phenylpropionaldehyde, TRC
Product Code. 30411780
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411780 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30411780 Supplier Toronto Research Chemicals Supplier No. TRCP33617510G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Phenylpropionaldehyde
CAS 93-53-8
Molecular Formula C9H10O
Quantity 10 g
Synonym alpha-Methyl-benzeneacetaldehyde, Hydratropaldehyde, (±)-2-Phenylpropanal, (±)-2-Phenylpropionaldehyde, (±)-Hydratropic Aldehyde, (±)-alpha-Phenylpropionaldehyde, 2-Phenylpropanal, 2-Phenylpropanaldehyde, Cumene Aldehyde, Hyacinthal, Hydratropic Aldehyde, NSC 5231, alpha-Formylethylbenzene, alpha-Methyl-alpha-toluic Aldehyde, alpha-Methylbenzeneacetaldehyde, alpha-Methylphenylacetaldehyde, alpha-Phenylpropionaldehyde
InChI Formula InChI=1S/C9H10O/c1-8(7-10)9-5-3-2-4-6-9/h2-8H,1H3
SMILES CC(C=O)c1ccccc1
IUPAC Name 2-phenylpropanal
Molecular Weight (g/mol) 134.18
Formula Weight 134.0732