missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropionaldehyde, 97%

Product Code. p-7026439
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460577 100 g 100g
11470577 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460577 Supplier Thermo Scientific Alfa Aesar Supplier No. B22436.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-53-8 | C9H10O | 134.178 g/mol

2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-53-8
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00006973
InChI Key IQVAERDLDAZARL-UHFFFAOYSA-N
Synonym 2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
PubChem CID 7146
IUPAC Name 2-phenylpropanal
SMILES CC(C=O)C1=CC=CC=C1

Specifications

Density 1.01
Boiling Point 92°C to 94°C (12 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.517
Quantity 100 g
Beilstein 4291321
Sensitivity Air Sensitive
Solubility Information Soluble in most fixed oils,propylene glycol. Insoluble in glycerin.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 2-Phenylpropionaldehyde
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.