Learn More
2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals
Product Code. 11456166
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456166 100 g 100g
11436166 5 g 5g
11446166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11456166 Supplier Thermo Scientific Alfa Aesar Supplier No. B22997.22
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36854-57-6
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00018811
InChI Key QGXMHCMPIAYMGT-UHFFFAOYSA-N
Synonym 2-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenylbutyrylchloride, butyryl chloride, 2-phenyl, acmc-209ipa, 2-phenyl butyryl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, .alpha.-phenylbutyryl chloride, 2-phenyl-2-ethylacetyl chloride
PubChem CID 98173
IUPAC Name 2-phenylbutanoyl chloride
SMILES CCC(C1=CC=CC=C1)C(=O)Cl
Specifications
Density 1.093
Boiling Point 143°C to 145°C (15 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.517
Quantity 100 g
UN Number UN3265
Beilstein 2045209
Sensitivity Moisture sensitive
Formula Weight 182.65
Percent Purity 98%
Chemical Name or Material 2-Phenylbutyryl chloride