Learn More
2-Oxo-3-hydroxy-N-methyl-N-propyl D-Lysergamide(Mixture of Diastereomers), TRC
Product Code. 30409341
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409341 1 mg
30427387 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30409341 Supplier Toronto Research Chemicals Supplier No. TRCO8568801MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 249921-57-1
Molecular Formula C20H25N3O3
Molecular Weight (g/mol) 355.44
Synonym (8beta)-9,10-Didehydro-2,3-dihydro-3-hydroxy-N,6-dimethyl-2-oxo-N-propylergoline-8-carboxamide, 2-Oxo-3-hydroxy LAMPA, 2-Oxo-3-hydroxylysergic Acid Methyl Propyl Amide
IUPAC Name (6aR,9R)-5a-hydroxy-N,7-dimethyl-5-oxo-N-propyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
SMILES CCCN(C)C(=O)[C@H]1CN(C)[C@@H]2CC3(O)C(=O)Nc4cccc(C2=C1)c34
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C20H25N3O3/c1-4-8-22(2)18(24)12-9-14-13-6-5-7-15-17(13)20(26,19(25)21-15)10-16(14)23(3)11-12/h5-7,9,12,16,26H,4,8,10-11H2,1-3H3,(H,21,25)/t12-,16-,20?/m1/s1
Formula Weight 355.1896
Chemical Name or Material 2-Oxo-3-hydroxy-N-methyl-N-propyl D-Lysergamide(Mixture of Diastereomers)