Learn More
2-(oxiran-2-yl)furan, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(oxiran-2-yl)furan |
| CAS | 2745-17-7 |
| Molecular Formula | C6H6O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C6H6O2/c1-2-5(7-3-1)6-4-8-6/h1-3,6H,4H2 |
| SMILES | C1OC1c2occc2 |
| IUPAC Name | 2-(oxiran-2-yl)furan |
| Molecular Weight (g/mol) | 110.11 |
| Formula Weight | 110.0368 |