Learn More
2-(Nonylthio)propanamide, TRC
Product Code. 30465241
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465241 50 mg 50mg
30470778 250 mg -
30471601 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30465241 Supplier Toronto Research Chemicals Supplier No. TRCN65048550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1021343-21-4
Molecular Formula C6 H10 O2
Molecular Weight (g/mol) 114.142
IUPAC Name 1-methylcyclobutane-1-carboxylic acid
SMILES CC1(CCC1)C(=O)O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H10O2/c1-6(5(7)8)3-2-4-6/h2-4H2,1H3,(H,7,8)
Formula Weight 114.068
Chemical Name or Material 2-(Nonylthio)propanamide