missing translation for 'onlineSavingsMsg'
Learn More

2-Nonanone, 98+%

Product Code. p-7081696
Change view
Click to view available options
Quantity:
50 g
100 g
1000 g
Unit Size:
1000g
100g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439353 50 g 50g
11309569 100 g 100g
11429353 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439353 Supplier Thermo Scientific Alfa Aesar Supplier No. A11781.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-55-6 | C9H18O | 142.242 g/mol

2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-55-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00009553
InChI Key VKCYHJWLYTUGCC-UHFFFAOYSA-N
Synonym 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
PubChem CID 13187
ChEBI CHEBI:77927
IUPAC Name nonan-2-one
SMILES CCCCCCCC(=O)C

Specifications

Melting Point -15°C
Density 0.826
Boiling Point 195°C
Flash Point 64°C (147°F)
Refractive Index 1.421
Quantity 50 g
Beilstein 1743645
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 142.24
Percent Purity ≥98%
Chemical Name or Material 2-Nonanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.