Learn More
Melford 2-Nitrophenyl-β-D-Xylopyranoside
Product Code. 30018596
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018596 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018596 Supplier Melford Supplier No. N638000.5
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Spectrophotometric substrate for detection of β-Xylosidase.
Spectrophotometric substrate for detection of β-Xylosidase. Protect from moisture.
Specifications
Quantity 500 mg