missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroanisole, 99%

Product Code. p-7024586
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454087 500 g 500g
11464087 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454087 Supplier Thermo Scientific Alfa Aesar Supplier No. A19518.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-23-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-23-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007096
InChI Key CFBYEGUGFPZCNF-UHFFFAOYSA-N
Synonym 2-nitroanisole, o-nitroanisole, anisole, o-nitro, 1-nitro-2-methoxybenzene, 2-methoxynitrobenzene, o-nitrophenyl methyl ether, benzene, 1-methoxy-2-nitro, 2-methoxy-1-nitrobenzene, benzene, methoxynitro, o-nitro methoxy benzene
PubChem CID 7048
ChEBI CHEBI:48722
IUPAC Name 1-methoxy-2-nitrobenzene
SMILES COC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 9°C to 10°C
Density 1.254
Boiling Point 272°C to 273°C
Flash Point 142°C (287°F)
Refractive Index 1.561
Quantity 500 g
UN Number UN2730
Beilstein 1868032
Merck Index 14,6585
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material 2-Nitroanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.