Learn More
2-Nitro Nevirapine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Nitro Nevirapine |
| CAS | 284686-16-4 |
| Molecular Formula | C15H13N5O3 |
| Quantity | 10 mg |
| Synonym | 11-Cyclopropyl-5,11-dihydro-4-methyl-2-nitro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| InChI Formula | InChI=1S/C15H13N5O3/c1-8-7-11(20(22)23)17-14-12(8)18-15(21)10-3-2-6-16-13(10)19(14)9-4-5-9/h2-3,6-7,9H,4-5H2,1H3,(H,18,21) |
| SMILES | Cc1cc(nc2N(C3CC3)c4ncccc4C(=O)Nc12)[N+](=O)[O-] |
| IUPAC Name | 11-cyclopropyl-4-methyl-2-nitro-5H-dipyrido[3,2-a:2',3'-d][1,4]diazepin-6-one |
| Molecular Weight (g/mol) | 311.3 |
| Formula Weight | 311.1018 |