Learn More
2-Nitro-N1-phenylbenzene-1,4-diamine, TRC
Specifications
Specifications
| CAS | 2784-89-6 |
| Molecular Formula | C12H11N3O2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C12H11N3O2/c13-9-6-7-11(12(8-9)15(16)17)14-10-4-2-1-3-5-10/h1-8,14H,13H2 |
| SMILES | Nc1ccc(Nc2ccccc2)c(c1)[N+](=O)[O-] |
| IUPAC Name | 2-nitro-1-N-phenylbenzene-1,4-diamine |
| Molecular Weight (g/mol) | 229.23 |
| Formula Weight | 229.0851 |
| Chemical Name or Material | 2-Nitro-N1-phenylbenzene-1,4-diamine |