Learn More
2-Nitro-N1-phenylbenzene-1,4-diamine, TRC
Product Code. 30179886
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179886 10 g 10g
30179752 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179886 Supplier Toronto Research Chemicals Supplier No. TRCN23272010G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2784-89-6
Molecular Formula C12H11N3O2
Molecular Weight (g/mol) 229.23
IUPAC Name 2-nitro-1-N-phenylbenzene-1,4-diamine
SMILES Nc1ccc(Nc2ccccc2)c(c1)[N+](=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C12H11N3O2/c13-9-6-7-11(12(8-9)15(16)17)14-10-4-2-1-3-5-10/h1-8,14H,13H2
Formula Weight 229.0851
Chemical Name or Material 2-Nitro-N1-phenylbenzene-1,4-diamine