Learn More
2-n-Butylfuran, 98%
Product Code. 11431013
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431013 250 g 250g
11411013 10 g 10g
11421013 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11431013 Supplier Thermo Scientific Alfa Aesar Supplier No. A13376.30
 Restricted Item
In Stock
Add to basket
CAS: 4466-24-4 | C8H12O | 124.183 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4466-24-4
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00047071
InChI Key NWZIYQNUCXUJJJ-UHFFFAOYSA-N
Synonym furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane
PubChem CID 20534
IUPAC Name 2-butylfuran
SMILES CCCCC1=CC=CO1
Specifications
Density 0.886
Boiling Point 153°C to 154°C
Flash Point 32°C (89°F)
Odor Pleasant
Quantity 250 g
UN Number UN1993
Beilstein 107045
Formula Weight 124.18
Percent Purity 98%
Chemical Name or Material 2-n-Butylfuran