Learn More
2-(Morpholinothio)benzothiazole, TRC
Product Code. 30471582
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471582 250 mg 250mg
30465194 500 mg 500mg
30482577 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30471582 Supplier Toronto Research Chemicals Supplier No. TRCM732783250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102-77-2
Molecular Formula C11H12N2OS2
Molecular Weight (g/mol) 252.35
Synonym Benzothiazole, 2-(4-morpholinylthio)- (ACI), 2-(4-Morpholinylthio)benzothiazole (ACI), Benzothiazole, 2-(morpholinothio)- (6CI, 7CI, 8CI), Morpholine, 4-(2-benzothiazolylthio)- (9CI), 2-(4-Morpholinothio)benzothiazole, 2-(4-Morpholinylmercapto)benzothiazole, 2-(Morpholin-4-ylsulfanyl)-1,3-benzothiazole, 2-(Morpholinothio)benzothiazole, 2-Benzothiazolesulfenemorpholide, 2-Benzothiazolesulfenic acid morpholide, 2-Benzothiazolyl N-morpholino sulfide, 2-Benzothiazolylsulfenylmorpholine, 4-(2-Benzothiazolylthio)morpholine, Accel NS, Accelerator NS, Amax, Cure-rite OBTS, Delac MOR, MBS, MBS 80, Meramid M, N,N-(Oxydiethylene)benzothiazole-2-sulfenamide, N-(Oxidiethylene)-2-benzothiazolyl sulfenamide, N-Morpholino-2-benzothiazolylsulfenamide, N-Oxydiethyl-2-benzothiazolesulfenamide, N-Oxydiethylene-2-benzothiazolylsulfenamide, NOBS, NOBS Special, Nocceler MSA, Nocceler MSA-G, NS, NSC 70078, OBTS, Pilcure MOR, Rhenogran MBS 80, SA-M, Sanceler NOB, Santocure MBS, Santocure MOR, Soxinol NBS, Sulfenamide M, Sulfenax MOR, Vulcafor BSM, Vulcafor MBS, Vulkacit MOZ, Vulkacit MOZ/LG
IUPAC Name 4-(1,3-benzothiazol-2-ylsulfanyl)morpholine
SMILES C1CN(CCO1)Sc2nc3ccccc3s2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C11H12N2OS2/c1-2-4-10-9(3-1)12-11(15-10)16-13-5-7-14-8-6-13/h1-4H,5-8H2
Formula Weight 252.0391
Chemical Name or Material 2-(Morpholinothio)benzothiazole