Learn More
2-Methyltryptamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyltryptamine Hydrochloride |
| CAS | 2826-95-1 |
| Molecular Formula | C11H14N2.HCl |
| Quantity | 250 mg |
| Synonym | 2-Methyl-1H-indole-3-ethanamine Hydrochloride, 3-(2-Aminoethyl)-2-methyl-indole Hydrochloride |
| InChI Formula | InChI=1S/C11H14N2.ClH/c1-8-9(6-7-12)10-4-2-3-5-11(10)13-8;/h2-5,13H,6-7,12H2,1H3;1H |
| SMILES | Cc1[nH]c2ccccc2c1CCN |
| Molecular Weight (g/mol) | 210.7 |