missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzonitrile, 98%

Product Code. p-7044511
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11357026

Brand: Thermo Scientific Alfa Aesar L06870.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6609-54-7 | C8H7NS | 149.211 g/mol

2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6609-54-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00015557
InChI Key PXZSANDJGNKIIA-UHFFFAOYSA-N
Synonym 2-methylthio benzonitrile, 2-methylsulfanyl benzonitrile, 2-cyanophenyl methyl sulfide, 2-methylmercapto benzonitrile, methylthio benzonitrile, 2-methylthiobenzenecarbonitrile, benzonitrile,2-methylthio, 2-methylsulfanyl benzenecarbonitrile
PubChem CID 138781
IUPAC Name 2-methylsulfanylbenzonitrile
SMILES CSC1=CC=CC=C1C#N

Specifications

Melting Point 33°C to 38°C
Boiling Point 116°C to 117°C (5 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 2207135
Solubility Information Insoluble in water.
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.