Learn More
2-(Methylsulfonyl)-5-(trifluoromethyl)-1,3,4-thiadiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Methylsulfonyl)-5-(trifluoromethyl)-1,3,4-thiadiazole |
| CAS | 27603-25-4 |
| Molecular Formula | C4H3F3N2O2S2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C4H3F3N2O2S2/c1-13(10,11)3-9-8-2(12-3)4(5,6)7/h1H3 |
| SMILES | CS(=O)(=O)c1nnc(s1)C(F)(F)F |
| IUPAC Name | 2-methylsulfonyl-5-(trifluoromethyl)-1,3,4-thiadiazole |
| Molecular Weight (g/mol) | 232.2 |
| Formula Weight | 231.9588 |